Behenic acid
Catalog # EHERC13057800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:340.59 g/mol
Merck Index:13,01024
Boiling Pt:306 °C (1013 hPa)
Melting Pt:80 °C
Density:0.8221 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-010-8
CAS Number:112-85-6
Formula:C₂₂H₄₄O₂
MDL Number:MFCD00002807