Order Entry
Norway
ContactUsLinkComponent
 
Isobutyl acetate for synthesis, Sigma-Aldrich®
 :  Merck
 
Isobutyl acetate for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    7 g/l (20 °C)
  • Melting Point:
    -99 °C
  • Molar Mass:
    116.16 g/mol
  • Boiling Point:
    116 - 118 °C (1013 hPa)
  • Vapor Pressure:
    17 hPa (20 °C)
  • Flash Point:
    18 °C
  • Refractive Index:
    1.3877 (20 °C, 589 nm)
  • Explosion Limit:
    2.4 - 10.5 %(V)
  • Density:
    0.87 g/cm3 (20 °C)
  • pH:
    5 (4 g/l, H2O, 20 °C)
  • Ignition Point:
    510 °C
  • Evaporation number:
    8