Order Entry
Norway
ContactUsLinkComponent
Acetone 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Flammable
Irritant
Health
Acetone 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Health
Acetone 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Meets E.P. Chemical Specifications:
  • Meets B.P. Chemical Specifications:
  • Meets N.F. Requirements:
  • GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Specific Gravity at 25°/25°C:
    ≤0.789
  • NF - Nonvolatile Residue:
    ≤0.004 %
  • NF - Readily Oxidizable Substances:
    Passes Test
  • NF - Water (H₂O):
    ≤0.5 %
  • NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis):
    99.5 - 100.5 %
  • NF - Benzene:
    ≤2 ppm
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Density (g/mL) at 20°C:
    0.790 - 0.793
  • EP/BP - Related Substances - Impurity A:
    ≤0.05 %
  • EP/BP - Related Substances - Impurity B:
    ≤0.05 %
  • EP/BP - Related Substances - Impurity C:
    ≤2 ppm
  • EP/BP- Other Impurities:
    ≤0.05 %
  • EP/BP - Matter Insoluble in H₂O:
    Passes Test
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Residue after Evaporation:
    ≤50 ppm
  • EP/BP - Water (H₂O), g/L:
    ≤3 g/l

Specification Test Results

Related Information

Frequently Bought Together