Order Entry
Norway
ContactUsLinkComponent
 
Pineapple Bromelain, MP Biomedicals
 
Pineapple Bromelain, MP Biomedicals

 

  • CAS No.:
    9001-00-7
  • Species:
    Pineapple
  • Storage conditions:
    0 °C
  • Form:
    Tan powder
  • Enzyme name:
    Bromelain
  • Enzyme activity:
    1100 - 1500 GDU/gram
  • pH:
    3,0 - 6,5