
(±)-Camphor-10-sulphonic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5872-08-2
:
Formula:C₁₀H₁₆O₄S
MDL Number:MFCD00074827
:232.3
Melting Pt:∼ 200 °C (dec.)
Storage Temperature:Ambient
EINECS:227-527-0
ADR:8,III
:GHS05