Daidzein
Catalog # MOLEM17127849
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.24 g/mol
Boiling Pt:513 °C (1013 hPa)
Melting Pt:212…214 °C
EINECS:207-635-4
CAS Number:486-66-8
Formula:C₁₅H₁₀O₄
MDL Number:MFCD00016954