Order Entry
Norway
ContactUsLinkComponent
Daidzein
Daidzein
Catalog # MOLEM17089343
Supplier:  Molekula
CAS Number:  
Daidzein
Catalog # MOLEM17089343
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :254.24 g/mol
Boiling Pt:513 °C (1013 hPa)
Melting Pt:212…214 °C
EINECS:207-635-4
CAS Number:486-66-8
Formula:C₁₅H₁₀O₄
MDL Number:MFCD00016954