
3,4,5-Trihydroxybenzamide 98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:618-73-5
:
Formula:C₇H₇NO₄
MDL Number:MFCD00014800
:169.14 g/mol
Boiling Pt:403 °C (1013 hPa)
Melting Pt:224...229 °C (dec., lit.)
:M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
EINECS:210-561-5
:GHS07
Specification Test Results
- Appearance (Colour):White to Beige and Faint Brown to Brown
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Carbon:48.5 - 51.0 %
- Nitrogen:8.0 - 8.5 %
- Purity (HPLC):≥97.5 %