2-Nitro-1-naphthol
Catalog # MAYBJFD00259.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:189.17 g/mol
Melting Pt:123…125 °C
EINECS:210-131-7
CAS Number:607-24-9
Formula:C₁₀H₇NO₃
MDL Number:MFCD00003956