4'-Ethoxyacetophenone
Katalog # MAYBDP00432.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.2 g/mol
Boiling Pt:83 °C (0.04 mmHg)
Boiling Pt:83 °C (0,04 mmHg)
Melting Pt:39 °C
EINECS:216-825-6
CAS Number:1676-63-7
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009095