Order Entry
Norway
ContactUsLinkComponent
2,4-Dinitro-1-naphthol
2,4-Dinitro-1-naphthol
Catalog # CHRO1B-351.0025
Supplier:  CHROMA
CAS Number:  
2,4-Dinitro-1-naphthol
Catalog # CHRO1B-351.0025
Supplier:  CHROMA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :234.17 g/mol
Boiling Pt:408 °C (1013 hPa)
Melting Pt:130…133 °C
EINECS:210-093-1
 :GHS07
Storage Temperature:Ambient
CAS Number:605-69-6
Formula:C₁₀H₆N₂O₅
MDL Number:MFCD00065352