2',4'-Difluoroacetophenone
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:364-83-0
:
Formula:C₈H₆F₂O
MDL Number:MFCD00151261
:156.13 g/mol
Boiling Pt:80 °C (1013 hPa)
Density:1,23 g/cm³ (20 °C)
EINECS:206-667-6
:GHS07
Storage Temperature:Ambient