Order Entry
Norway
ContactUsLinkComponent
 
Tyramine 97%
 
undefined
Tyramine 97%

 

  • Appearance:
    Off-white to beige to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    158 to 163 °C
  • Titration with HClO4:
    ≥96.0 %