9,10-Dihydroanthracene
Catalog # MOLE45359989-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:180.25 g/mol
Boiling Pt:312 °C (1013 hPa)
Melting Pt:70…72 °C
Density:1,207 g/cm³ (20 °C)
EINECS:210-336-1
CAS Number:613-31-0
Formula:C₁₄H₁₂
MDL Number:MFCD00001239