Tin(II) chloride dihydrate, Supelco®
Catalog # SUPL96527-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
:GHS05
:GHS07
CAS Number:10025-69-1
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863