Tin(II) chloride dihydrate 98.0-103.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:14,8783
Boiling Pt:623 °C (1013 hPa)
Melting Pt:43…46 °C
Density:2.71
Storage Temperature:Ambient
EINECS:600-045-1
UN:3260
ADR:8,III
:GHS05
:GHS07