Tin(II) chloride dihydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:37…38 °C
Density:2,71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
:GHS07
:GHS08
:GHS05
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals and/or chunks
- Titration Iodimetric:≥96.0 %