
Citric acid monohydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:5949-29-1
:
:C₆H₈O₇.H₂O
:MFCD00149972
:210.14 g/mol
:13,02350
:135…152 °C
:1.552 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:173.9 °C (closed cup)
:201-069-1
:GHS07
:Ambient
- Assay (acidimetric, calc. on anhydrous substance):99.5 - 100.5 %
- Identity (IR-spectrum):passes test
- Appearance:white to almost white or colorless crystals
- Appearance of solution (200 g/l; water):clear (≤ 3 NTU) and not more intense in color than reference solution Y₇, BY₇ or GY₇
- In water insoluble matter:≤ 50 ppm
- Chloride (Cl):≤ 5 ppm
- Phosphate (PO₄):≤ 10 ppm
- Sulphate (SO₄):≤ 20 ppm
- Heavy metals (as Pb):≤ 5 ppm
- Al (Aluminium):≤ 0.2 ppm
- As (Arsenic):≤ 1 ppm
- Hg (Mercury):≤ 1 ppm
- Cu (Copper):≤ 5 ppm
- Fe (Iron):≤ 3 ppm
- Fe (Iron, acc. to Reag. Ph Eur.):suitable to use in the limit test for iron
- Pb (Lead):≤ 0.5 ppm
- Oxalates (as C₂H₂O₄):≤ 50 ppm
- Readily carbonisable substance:passes test
- Sulfated ash:≤ 200 ppm
- Water (according to Karl Fischer):7.5 - 8.8 %