Order Entry
Norway
ContactUsLinkComponent
Nitric acid concentrate for dilution to 0.1 mol/l in aqueous solution, Titrisol®, Supelco®
Corrosive
Nitric acid concentrate for dilution to 0.1 mol/l in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09964.0001
Supplier:  Merck
CAS Number:  
Corrosive
Nitric acid concentrate for dilution to 0.1 mol/l in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09964.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -10 °C
  • Boiling Point:
    101 °C (1013 hPa)
  • Density:
    1.07 g/cm3 (20 °C)
  • pH:
     - 1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :63.01 g/mol
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:231-714-2
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349