Sodium chloride 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
1.09945.0001
: Merck
:
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:0501
- Solubility:(20 °C) soluble
- Density:1.08 g/cm3 (20 °C)
- pH:4.8 (H2O, 20 °C)
:
:58.44 g/mol
:M-Clarity™ quality level = MQ100
:Ambient
:231-598-3
:7647-14-5
:NaCl
:MFCD00003477