Order Entry
Norway
Orders LinkContactUsLinkComponent
 

(±)-2-Octanol

Glass bottle for solids, 1 g
Catalog # ROTH9439.1
Flammable
Irritant
Marketsource
Supplier:   Roth Carl
(±)-2-Octanol
Catalog # ROTH9439.1
Supplier:  Roth Carl
Marketsource
CAS Number:  123-96-6
(±)-2-Octanol

Glass bottle for solids, 1 g

CAS Number:  123-96-6
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
  • CAS Number:123-96-6
  • Formula:C₈H₁₈O
  • MDL Number:MFCD00004591
  • Molecular Weight:130.23 g/mol
  • Merck Index:13,06785
  • Boiling Pt:180 °C (1013 hPa)
  • Melting Pt:–39 °C
  • Density:0.818 g/cm³ (20 °C)
  • Flash Pt:55 °C
  • Storage Temperature:Ambient
  • EINECS:204-667-0
  • UN:1987
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications