Order Entry
Norway
Orders LinkContactUsLinkComponent
(±)-2-Octanol ≥98%

(±)-2-Octanol ≥98%

(±)-2-Octanol ≥98%
Certificates
(±)-2-Octanol ≥98%
(±)-2-Octanol ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 123-96-6 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 76°(169°F) Refractive Index: 1.4260 Odour: Pleasant Melting Point: -38° Boiling Point: 179-180° Beilstein Reference: 1719325 Merck Reference: 14,6752 MDL No.: MFCD00004591

Synonyms:  (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
  • CAS Number:123-96-6
  • Formula:C₈H₁₈O
  • MDL Number:MFCD00004591
  • Molecular Weight:130.23 g/mol
  • Merck Index:14,6752
  • Boiling Pt:179…180 °C
  • Melting Pt:–38 °C
  • Density:0.819
  • Flash Pt:76 °C (169 °F)
  • Storage Temperature:Ambient
  • EINECS:204-667-0
  • UN:1987
  • ADR:3,III
Product Family Options

Product Information