(±)-2-Octanol 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
- CAS Number:123-96-6
- Formula:C₈H₁₈O
- MDL Number:MFCD00004591
- Molecular Weight:130.23 g/mol
- Merck Index:13,06785
- Boiling Pt:174…181 °C (760 mmHg)
- Melting Pt:–38 °C
- Density:0.81 g/cm³
- Flash Pt:72 °C
- Storage Temperature:Ambient
- EINECS:204-667-0
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
- CAS Number:123-96-6
- Formula:C₈H₁₈O
- MDL Number:MFCD00004591
- Molecular Weight:130.23 g/mol
- Merck Index:13,06785
- Boiling Pt:174…181 °C (760 mmHg)
- Melting Pt:–38 °C
- Density:0.81 g/cm³
- Flash Pt:72 °C
- Storage Temperature:Ambient
- EINECS:204-667-0
- ADR:6.1,III
Documents
ACRO12941
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4270 (20°C, 589 nm)
- GC:≥96.0 %
- Titration with NaOH:≥98.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



