Order Entry
Norway
Orders LinkContactUsLinkComponent
Hydrogen peroxide 35%

Hydrogen peroxide 35%

5 L
Catalog # SWED17477-5
Corrosive
Irritant
Supplier:   SWED HANDLING
Hydrogen peroxide 35%
Catalog # SWED17477-5
Supplier:  SWED HANDLING
CAS Number:  7722-84-1
Hydrogen peroxide 35%

5 L

CAS Number:  7722-84-1
Price
kr 575,00
Log in for availability
1 * 5 L
Product Details & Documents
  • CAS Number:7722-84-1
  • Formula:H₂O₂
  • MDL Number:MFCD00011333
  • Molecular Weight:34.01 g/mol
  • Boiling Pt:∼107 °C (1013 hPa)
  • Melting Pt:< 0 °C
  • Density:1.11 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-765-0
  • UN:2014
  • ADR:5.1,II
  •  :Density: 1.11 g/cm³ (20 °C)
Specifications

Specifications

  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.