Hydrofluoric acid 48%, AnalaR NORMAPUR® analytical reagent
Katalog # 20319.291
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 48 %
- Colouration:Max. 10 APHA
- SiF6 (Hexafluorosilicate):Max. 100 ppm
- PO4 (Phosphate):Max. 0.5 ppm
- SO4 + SO3 (as SO4):Max. 5 ppm
- Ag (Silver):Max. 0.02 ppm
- Al (Aluminium):Max. 0.1 ppm
- As (Arsenic):Max. 0.05 ppm
- Ba (Barium):Max. 0.1 ppm
- Bi (Bismuth):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Cl (Chloride):Max. 5 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.3 ppm
- K (Potassium):Max. 0.1 ppm
- Li (Lithium):Max. 0.02 ppm
- Mg (Magnesium):Max. 0.2 ppm
- Mn (Manganese):Max. 0.05 ppm
- Mo (Molybdenum):Max. 0.05 ppm
- Na (Sodium):Max. 0.2 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Sr (Strontium):Max. 0.02 ppm
- Ti (Titanium):Max. 0.1 ppm
- V (Vanadium):Max. 0.05 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
About this item
:
:20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8 (6.1),II
:GHS06
:GHS05
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346