Hydrogen peroxide 30% stabilised
Plastic bottle, 250 mL
Catalog # ROTH8070.2
Marketsource
Supplier: Roth Carl
Hydrogen peroxide 30% stabilised
Catalog # ROTH8070.2
Supplier: Roth Carl
Marketsource
CAS Number: 7722-84-1
Jump to:
Product Details & Documents
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:∼107 °C (1013 hPa)
- Melting Pt:< 0 °C
- Density:1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
- UN:2014
- ADR:5.1,II
- :Density: 1.11 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



