1,3-Dimethylcyclohexane
Catalog # EHERCA12726430
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:112.22 g/mol
Boiling Pt:120 °C (1013 hPa)
Density:0.7672 g/cm³ (20 °C)
EINECS:209-707-0
UN:2263
ADR:3,II
CAS Number:591-21-9
Formula:C₈H₁₆
MDL Number:MFCD00064173