1,3-Dichloropropene-[D₄] (98% D)
Catalog # PROOCILDLM19380.1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
UN:2047
ADR:3,III
:GHS02
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:202656-23-3
Formula:C₃D₄Cl₂