Cleanroom overalls, Kimtech™ A5
About this item
Kimtech™ A5 sterile cleanroom apparel with CLEAN-DON™ technology is made of SMS (Spunbond Melt-blown Spunbond) material. It features built-in snaps that gather up legs and arms to aid aseptic gowning. Garments have a thumb loop that prevents the sleeve from rolling back and exposing arm or wrist when in use.
- Cloth-like comfort
- Innovative inside-out fold pattern - reduces the risk of touching and contaminating the outside of the apparel when opening or donning
- Highly visible blue line along the inside of the garment signals the proper place to grasp while gowning
- Vacuum packaging reduces space, so more products fit into gowning area storage
Suitable for use in Class 100, ISO 5, 3.5 environments.
Colour: White
: PPE Cat III (EU) 2016/425, Type 5/6
: Vacuum packed and double-bagged for added sterility assurance.
7 Options Available Below
Product Details & Documents
Kimtech™ A5 sterile cleanroom apparel with CLEAN-DON™ technology is made of SMS (Spunbond Melt-blown Spunbond) material. It features built-in snaps that gather up legs and arms to aid aseptic gowning. Garments have a thumb loop that prevents the sleeve from rolling back and exposing arm or wrist when in use.
- Cloth-like comfort
- Innovative inside-out fold pattern - reduces the risk of touching and contaminating the outside of the apparel when opening or donning
- Highly visible blue line along the inside of the garment signals the proper place to grasp while gowning
- Vacuum packaging reduces space, so more products fit into gowning area storage
Suitable for use in Class 100, ISO 5, 3.5 environments.
Colour: White
: PPE Cat III (EU) 2016/425, Type 5/6
: Vacuum packed and double-bagged for added sterility assurance.
Specifications for Product Family
Specifications
- Catalog No:
- 113-1198
- 113-1199
- 113-1210
- 113-1211
- 113-1212
- 113-1213
- 113-1219
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
Specifications
Product Family Options
Product Information
- ColourClothing SizeSterileAvailabilityPrice
- 113-1198Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1198Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing - 113-1199Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1199Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing - 113-1210Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1210Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing - 113-1211Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1211Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing - 113-1212Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1212Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing - 113-1213Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1213Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing - 113-1219Without hoodSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1219Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWaste Reducing
Complementary Products
Product Information
- Catalog #DescriptionShoe sizeSterileColourLengthAvailabilityPrice