Iodine 99.5-100.5% Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:13,05036
Boiling Pt:184.3 °C (1013 hPa)
Melting Pt:114 °C
Density:4.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8 (6.1),III
REACH:01-2119485285-30
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay:99.5 - 100.5%
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes Test
- Identification B:Passes Test
- Bromides and Chlorides:Max. 250 ppm
- Non-Volatile Residue:Max. 0.1%
- Residual Solvents:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC24762.265 spec_VWRC24762.265 spec_VWRC24762.290 spec_VWRC24762.290 spec_VWRC24762.290
Product specification
VWR Chemicals for Biopharma and Pharma Production