kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Boron
Bor ≥98% (metals basis), crystalline powder -325 mesh
Bor ≥98% (metals basis), crystalline powder -325 mesh
:
Thermo Fisher Scientific
Bor ≥98% (metals basis), crystalline powder -325 mesh
:
Thermo Fisher Scientific
:
7440-42-8
:
:
B
:
MFCD00134034
:
10.81 g/mol
:
13,01333
:
Romtemperatur
:
231-151-2
:
3178
:
4.1,III
:
GHS02
:
GHS07