Centrifuge tubes, EZ Flip™, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP, conical, printed graduations with hinged cap.
- Printed graduations, white writing area and flat caps for sample identification
- Hinged-cap system with ergonomic design for one handed opening and closing
- Integrated leakproof cap prevents cross-contamination and lost closures
- Withstands forces up to 8500 ×g for 15 ml and 9500 ×g for 50 ml
These tubes are manufactured from high grade polypropylene. RNase-/DNase-free and non pyrogenic.
Packaging: Bagged tubes are packed in durable and re-sealed (zip closure) bags.
Specifications
- Catalog No:
- NUNC362696NS
- NUNC362695NS
- NUNC362694NS
- NUNC362697NS
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement