Capsaicin
Catalog # EHERC10949000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:305.42 g/mol
Merck Index:12,01811
Boiling Pt:210…220 °C (0.01 mmHg)
Melting Pt:66.1 °C
EINECS:206-969-8
UN:2811
ADR:6.1,III
CAS Number:404-86-4
Formula:C₁₈H₂₇NO₃
MDL Number:MFCD00017259