Natriumhydroksid 3% vandig løsning
Catalog # LBSE790913
Supplier: LABSERVICE
CAS Number:
Specifications
- Fp:5 L
- Seq:5190
Specifications
About this item
:
:40 g/mol
Storage Temperature:Romtemperatur
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548