Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bismarck brown Y
Bismarck brown Y, dark brown powder C.I. 21000
Bismarck brown Y, dark brown powder C.I. 21000
:
MP Biomedicals
Bismarck brown Y, dark brown powder C.I. 21000
:
MP Biomedicals
:
10114-58-6
:
:
C₁₈H₁₈N8·2HCl
:
MFCD00012977
:
419.32 g/mol
:
12,01294
:
Romtemperatur
:
233-314-3
Identity Test:
Passes
UV/Vis: l max :
452 - 457 nm (50% EtOH, few drops of 1N HCl)