Bismarck brown Y
Catalog # MOLEM61849094
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:419.32 g/mol
Merck Index:12,01294
Storage Temperature:Ambient
EINECS:233-314-3
CAS Number:10114-58-6
Formula:C₁₈H₁₈N8·2HCl
MDL Number:MFCD00012977