Order Entry
Norway
ContactUsLinkComponent
Bismarck brown R
Bismarck brown R
Catalog # CHRO1B-261
Supplier:  CHROMA
CAS Number:  
Bismarck brown R
Catalog # CHRO1B-261
Supplier:  CHROMA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :461.39 g/mol
Storage Temperature:Ambient
EINECS:213-888-1
CAS Number:5421-66-9
 :1052-38-6
Formula:C₂₁H₂₄N₈·2HCl
MDL Number:MFCD00036386