Bismarck brown R
Catalog # MOLEM38973565
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:461.39 g/mol
Storage Temperature:Ambient
EINECS:213-888-1
CAS Number:5421-66-9
:1052-38-6
Formula:C₂₁H₂₄N₈·2HCl
MDL Number:MFCD00036386