2,6-Dichlorophenolindophenol sodium salt ≥97.0% (by argentometric analysis, on dry substance), Fluka™
:620-45-1
:
:C₁₂H₆Cl₂NNaO₂
:290.08 g/mol
:13,03093
:>300 °C
:Ambient
:210-640-4
- assay (HPLC):Min. 99 %
- measurement uncertainty:effective %
- identity (NMR):complying
- melting range:124 - 127 °C
- Identity (IR):complying
- water (Karl Fischer):Max. 5 %
Frequently Bought Together