

Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:28300-74-5
:304-61-0
Formula:C₈H₄K₂O₁₂Sb₂·3H₂O
MDL Number:MFCD00167056
:667.88 g/mol
Merck Index:14,700
Melting Pt:100 °C
Density:2.6
Storage Temperature:Ambient
EINECS:229-436-1
UN:1551
ADR:6.1,III
:GHS07
:GHS09