Order Entry
Norway
ContactUsLinkComponent
Carbol fuchsin (Ziehl-Neelsen)
Carbol fuchsin (Ziehl-Neelsen)
Catalog # LBSE791181
Supplier:  LABSERVICE
CAS Number:  
Carbol fuchsin (Ziehl-Neelsen)
Catalog # LBSE791181
Supplier:  LABSERVICE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :351.88 g/mol
Density:0.99 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-189-6
UN:1992
ADR:3,III
 :GHS02
 :GHS05
 :GHS08
 :GHS07
CAS Number:4197-24-4
Formula:C₂₁H₂₂ClN₃
MDL Number:MFCD18071399