Order Entry
Norway
ContactUsLinkComponent
L(-)-Menthol, Sigma-Aldrich®
L(-)-Menthol, Sigma-Aldrich®
Catalog # 8.14113.0100
Supplier:  Merck
CAS Number:  
L(-)-Menthol, Sigma-Aldrich®
Catalog # 8.14113.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    490 mg/l (25 °C)
  • Melting Point:
    41 - 44 °C
  • Molar Mass:
    156.26 g/mol
  • Bulk Density:
    450 kg/m3
  • Boiling Point:
    216 °C
  • Flash Point:
    101 °C
  • Density:
    0.890 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :156.27 g/mol
Merck Index:13,05861
Boiling Pt:212 °C (1013 hPa)
Melting Pt:41…44 °C
Density:0.89 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:218-690-9
 :GHS07
CAS Number:2216-51-5
Formula:C₁₀H₂₀O
MDL Number:MFCD00062979