Stirrer Glands, PFTE or Brass, Quickfit®
Supplier: SciLabware
Certificates
About this item
Stuffing box type glands with choice of brass or PTFE cones. Accepts 6 mm diameter stirrer shaft.
- Provides a seal by means of adjustable compression of O rings and lubricated felt washers
- 201-8730 and 201-8731 have integral drive pulleys