About this item
Peristaltic pump heads SP standard 1,6 mm and SP standard 2,5 mm: For simple pumping tasks. Housing made from die-cast aluminium. Rollers made from stainless steel, roller bearings made from polyamide. Rotor equipped with 2 convex rollers; cell cultures will not be damaged. For tubing with wall thickness of 1,6 or 2,5 mm. Rated flow: 0,5 to 3644 ml/min (depending on the drive and tubing used).
Peristaltic pump head SP vario: For tubing with various wall thicknesses
Housing made from die-cast aluminium
Rollers made from stainless steel, roller bearings made from coated aluminium
Rotor equipped with 2 convex rollers, cell cultures will not be damaged
Tubing with wall thicknesses of 1,6 and 2,5 mm; rotor with adjustable roller clearance to allow for the thickness of the tubing wall
Flow rate: 0,5 to 3644 ml/min (depending on the drive and tubing used)
Specifications
- Catalog No:
- 224-1362
- HEID523-43020-00
- Min. flow rate:
- 0,5 ml/min
- 0,5 ml/min