Single channel pipettes, mechanical, fixed / variable volume, Proline®
Ergonomic pipettes with fast, easy operation. Selected models offer changeable tip cone filters to prevent liquids and aerosols entering the pipette.
- Easy calibration and maintenance, small number of parts
- Smooth plunger action and simple in-lab recalibration and service
- Volume setting by a simple click-stop
- Ergonomic finger support, which reduces the risk of RSI
Two types of filters are offered: Safe-Cone standard filter and Safe-Cone Plus filter. It is recommended to use the Standard filter for general applications and the Plus filter for more demanding applications, such as cell culture, bacterial, viral work and molecular biology.
: Supplied with five extra filters, forceps and a calibration tool.
- Catalog No:
- 613-4087
- 613-4054
- 613-4088
- 613-4077
- 613-4078
- 613-4079
- 613-4045
- 613-4080
- 613-4053
- 613-4046
- 613-4081
- 613-4047
- 613-4082
- 613-4050
- 613-4083
- 613-4052
- 613-4084
- 613-4085
- 613-4051
- 613-4086
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001