Incubators, thermostatically controlled
Supplier: Lovibond
Certificates
About this item
The CFC-free thermostatically controlled incubators are designed to ensure continuous temperature control in a wide range of applications, such as: 20 °C BOD determination; 4 °C storage of wastewater samples; 25 °C enzyme activity testing (TTC-test); 37 °C colony counting.
- Fully insulated cabinet with universal temperature control unit
- Reliable temperature control within a range specified from 2 to 40 °C in steps of 1 °C
- Temperature tolerance ±1 °C, for a stirred 500 ml water test in the centre of the incubator at 20 °C
- LED with actual and target value, three digits, resolution 0,1 °C
- Choice of standard door or glass insulated glass door
Temperature range: ambient 2 to 40 °C.
Caution: * Suitable for under bench applications
* 600×600×820 (mm) without work top
* 600×600×850 (mm) with work top
* 600×600×820 (mm) without work top
* 600×600×850 (mm) with work top