Weighing table for micro and analytical balances
Supplier: Sartorius Balances
Jump to:
About this item
High performance micro and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast stabilisation of the reading. The YWT09 weighing table features a sturdy frame with a granite stone slab insert.
- Easy to clean surfaces
- Fast stabilisation times
Ordering information: Assembly is not included.
2 Options Available Below
Product Details & Documents
High performance micro and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast stabilisation of the reading. The YWT09 weighing table features a sturdy frame with a granite stone slab insert.
- Easy to clean surfaces
- Fast stabilisation times
Ordering information: Assembly is not included.
Product Family Options
Product Information
- W×D×HWeightAvailabilityPrice
- 611-1131YWT03 balance table made of cast stone with vibration dampersYWT03 balance table made of cast stone with vibration dampers900×600×760 mm238Log in for availability1 * 1 Itemskr 31 850,00
- 611-1702YWT09 balance table, wooden frame with cast stone slab insertYWT09 balance table, wooden frame with cast stone slab insert900×750×750 mmLog in for availability1 * 1 Itemskr 29 275,00