Mini horizontal electrophoresis systems, Owl™ B1 EasyCast™
Katalog # 730-0188
Supplier: THERMO OWL SCIENTIFIC
Specifications
- Model:Owl B1-BP
- Description:Mini horizontal gel system with buffer exchange ports
- Buffer volume:600 ml
- Gel size W×L:90×110 mm
- Unit W×L×H:150×220×95 mm
- Pk:1
Specifications
About this item
The durable, easy‑to‑use, Owl™ B1 EasyCast™ mini horizontal gel system is suitable for running between five and 28 samples and is ideal for most teaching applications.
- Two comb slots on the UV transparent gel tray double's sample capacity by doubling the number of sample wells
- Heavy duty, one-piece combs eliminate comb assembly
- UVT gel trays are silk screened with a fluorescent ruler for easy measurement of bands
- Available with optional buffer exchange ports for use with an external recirculation pump
Delivery: Each system includes buffer chamber, SuperSafe™ lid with attached power leads, gasketed UVT gel tray and two combs (10- and 14-well, double-sided, 1,0/1,5 mm thick).