Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Sensolyt® 690-7 assembly
Sensolyt® 690-7 assembly
Sensolyt® 690-7 assembly
Catalog # 708-0039
Supplier:
WTW ONLINE
Utvid bildet
Sensolyt® 690-7 assembly
Catalog # 708-0039
Supplier:
WTW ONLINE
Specifications
Description:
Sensolyt® 690-7 Assembly with built-in NTC sensor
Pk:
1
Specifications
About this item
Produktinformasjon
Produktinformasjon