Order Entry
Norway
ContactUsLinkComponent
VWR®, Accessories for Digital Block Heaters
VWR®, Accessories for Digital Block Heaters
Catalog # 460-3288
Supplier:  VWR Collection
undefined
VWR®, Accessories for Digital Block Heaters
Catalog # 460-3288
Supplier:  VWR Collection
Specifications
  • Description:
    External temperature probe kit. Kit includes stainless steel RTD probe, 457 mm vertical support rod, thermometer/probe extension clamp and hook connector.
  • Seq:
    098
  • Pk:
    1